Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MEHGSIITQA RREDALVLTK QGLVSKSSPK KPRGRNIFKA LFCCFRAQHV GQSSSSTELA AYKEEANTIA KSDLLQCLQY QFYQIPGTCL LPEVTEEDQ
CTDSP2 is one of several small CTD phosphatases (SCP). It has been proposed that SCP activity is an evolutionarily conserved transcriptional regulator that acts globally to silence neuronal genes. In addition, expression of phosphatase CTDSP2 mRNA isenhanced in several cancer cell lines with amplification of the CTDSP2 and CDK4 genes, and CTDSP2 coamplifies with CDK4 in some primary sarcomas. CTDSP2 may contribute to the development of a subset of sarcomas.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Carboxy-terminal domain RNA polymerase II polypeptide A small phosphatase 2; conserved gene amplified in osteosarcoma; CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase 2; NLI-interacting factor 2; Nuclear LIM interactor-interacting factor 2; Protein OS-4; SCP2; Small C-terminal domain phosphatase 2; Small CTD phosphatase 2
Gene Aliases: CTDSP2; NIF2; OS4; PSR2; SCP2
UniProt ID: (Human) O14595
Entrez Gene ID: (Human) 10106
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support