Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Probable phosphatase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase like 2; CTD small phosphatase-like protein 2; CTDSP-like 2
Gene Aliases: CTDSPL2; D2Ertd485e; HSPC058; HSPC129; RGD1309219
UniProt ID: (Human) Q05D32, (Rat) Q5XIK8, (Mouse) Q8BG15
Entrez Gene ID: (Human) 51496, (Rat) 311368, (Mouse) 329506
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support