Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bethyl Laboratories
Promotions
View available promotion(s)
Promo Code: CI2Z9E .
FIGURE: 1 / 3
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Monkey and Gorilla
CHTF18, CHTF8.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C321D2.3 (novel protein); C321D2.4 (novel protein); CHL12; Chromosome transmission fidelity protein 18 homolog; CTF18, chromosome transmission fidelity factor 18 homolog; hCTF18; homolog of yeast CHL12; some homology with holliday junction DNA helicase RUVB like
Gene Aliases: C16orf41; C321D2.2; C321D2.3; C321D2.4; CHL12; CHTF18; CTF18; RUVBL
UniProt ID: (Human) Q8WVB6
Entrez Gene ID: (Human) 63922
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support