Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: YLNARSSRDF EYYKQFCVPG FKNNKGVAEV LRDGDCPAVL IPSKPLARRC FPAIHAYKGV LMVGNETTYE DGHGSRKNIT DLVEGAKKAN GVLEARQLAM RIFEDYTV
Isoform 1, but not isoform 3, exhibits some choline transporter activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Choline transporter-like protein 2; solute carrier family 44 (choline transporter), member 2; Solute carrier family 44 member 2; solute carrier family 44, member 2; testicular tissue protein Li 47
Gene Aliases: CTL2; PP1292; PSEC0210; SLC44A2
UniProt ID: (Human) Q8IWA5
Entrez Gene ID: (Human) 57153
Molecular Function:
transporter
secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support