Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGSRGQGLL LAYCLLLAFA SGLVLSRVPH VQGEQQEWEG TEELPSPPDH AERAEEQHEK YRPSQDQGLP ASRCLRCCDP GTSMYPATAV PQINITILKG EKGDRGDRGL QGKYGKTGSA GARGHTGPKG QKGSMGAPGE RCKSHYAAFS VGRKKPMHSN HYYQTVIFDT EFVNLYDHFN MFTGKFYCYV PGLYFFSLNV HTWNQKETYL HIMKNEEEVV ILFAQVGDRS IMQSQSLMLE LREQDQVWVR LYKGERENAI FSEELDTYIT FSGYLVKHAT EP (1-281 aa encoded by BC021553)
Adipose tissue of an organism plays a major role in regulating physiologic and pathologic processes such as metabolism and immunity by producing and secreting a variety of bioactive molecules termed adipokines. One highly conserved family of adipokines is adiponectin/ACRP30 and its structural and functional paralogs, the C1q/tumor necrosis factor-alpha-related proteins (CTRPs) 1-7. Unlike adiponectin, which is expressed exclusively by differentiated adipocytes, the CTRPs are expressed in a wide variety of tissues. These proteins are thought to act mainly on liver and muscle tissue to control glucose and lipid metabolism. An analysis of the crystal structure of adiponectin revealed a structural and evolutionary link between TNF and C1q-containing proteins, suggesting that these proteins arose from a common ancestral innate immunity gene. In obese (ob/ob) mice, RT-PCR analysis showed that mCTRP1 transcripts are seen at substantially higher levels in adipose tissues compared to those of normal mice.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Complement C1q tumor necrosis factor-related protein 1; G protein coupled receptor interacting protein; G protein-coupled receptor-interacting protein; GIP
Gene Aliases: C1QTNF1; CTRP1; GIP; UNQ310/PRO353; ZSIG37
UniProt ID: (Human) Q9BXJ1
Entrez Gene ID: (Human) 114897
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support