Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: MNGTLDHPDQ PDLDAIKMFV GQVPRTWSEK DLRELFEQYG AVYEINVLRD RSQNPPQSKG CCFVTFYTRK AALEAQNALH NMKVLPGMHH PIQMKPADSE KNNAVEDRKL FIGMISKKCT ENDIRVMFSS FGQIEECRIL RGPDGLSRGC AFVTFTTRAM AQTAIKAMHQ AQTMEGCSSP MVVKFADTQK DKEQKRMAQQ LQQQMQQISA ASVWGNLAGL NTLGPQYLAL LQQTASSGNL NTLSSLHPMG GLNAMQLQNL AALAAAASAA QNTPSGTNAL TTSSSPLSVL TSSAGSSPSS SSSNSVNPIA SLGALQTLAG ATAGLNVGSL AGMAALNGGL GSSGLSNGTG STMEALTQAY SGIQQYAAAA LPTLYNQNLL TQQSIGAAGS QKEGPEGANL FIYHLPQEFG DQDLLQMFMP FGNVVSAKVF IDKQTNLSKC FGFVSYDNPV SAQAAIQSMN GFQIGMKRLK VQLKRSKNDS KPY
Myotonic dystrophy (MD) is an autosomal dominant neuromuscular disease that is associated with a (CTG)n repeat expansion in the 3'-untranslated region of the myotonin protein kinase (Mt-PK) gene. A (CUG) n oligonucleotides triplet repeat pre-mRNA/mRNA binding protein may play an important role in DM pathogenesis. HeLa cell protein, CUG-BP1, has been purified based upon its ability to bind specifically to (CUG) 8 oligonucleotides in vitro. CUG-BP1 is the major (CUG) 8 binding activity in normal cells. CUG-BP1 has been identified as isoforms of a novel heterogeneous nuclear ribonucleoprotein (hnRNP), hNab50. The CUG-BP/hNab50 protein is localized predominantly in the nucleus and is associated with polyadenylated RNAs in vivo. In vitro RNA-binding/photocrosslinking studies demonstrate that CUG-BP/hNab50 binds to RNAs containing the Mt-PK 3-UTR. The (CUG) n repeat region in Mt-PK mRNA is a binding site for CUG-BP/hNab50 in vivo, and triplet repeat expansion leads to sequestration of this hnRNP on mutant Mt-PK transcripts.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 50 kDa nuclear polyadenylated RNA-binding protein; Brain protein F41; bruno-like 2; Bruno-like protein 2; CELF-1; CUG RNA-binding protein; CUG triplet repeat RNA-binding protein 1; CUG triplet repeat, RNA binding protein 1; CUG triplet repeat, RNA-binding protein 1; CUG-BP- and ETR-3-like factor 1; CUG-BP1; CUGBP Ela; CUGBP Elav-like family member 1; Deadenylation factor CUG-BP; Deadenylation factor EDEN-BP; EDEN-BP homolog; embryo deadenylation element binding protein; Embryo deadenylation element-binding protein homolog; nuclear polyadenylated RNA-binding protein, 50-kD; RNA-binding protein BRUNOL-2; RP23-147D3.7
Gene Aliases: 1600010O03Rik; AA407467; BRUNOL2; CELF1; CUG-BP; CUG-BP1; CUGBP; CUGBP1; D2Wsu101e; EDEN-BP; HNAB50; NAB50; NAPOR
UniProt ID: (Human) Q92879, (Mouse) P28659
Entrez Gene ID: (Human) 10658, (Mouse) 13046
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support