Save $650 & get a lab-themed hidden object puzzle

Save and claim

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • CUX1/Protein CASP Antibodies

            Proteintech

            CUX1/Protein CASP Polyclonal Antibody

            47 References
            View all (6) CUX1/Protein CASP antibodies
            Datasheet
            Protocols
            Questions & Answers
            Promotion Available
            Learn more
            Catalog # 11733-1-AP
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            492.00
            150 µL
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite CUX1/Protein CASP Polyclonal Antibody

            • Antibody Testing Data (15)
            Application
            CUX1/Protein CASP Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 15

            CUX1/Protein CASP Antibody (11733-1-AP) in ICC/IF

            Immunofluorescent analysis of HepG2 cells, using CUX1 antibody 11733-1-AP at 1:25 dilution and Rhodamine-labeled goat anti-rabbit IGG (red).
            View Product

            CUX1/Protein CASP Antibody in Immunocytochemistry (ICC/IF)
            CUX1/Protein CASP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            CUX1/Protein CASP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            CUX1/Protein CASP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            CUX1/Protein CASP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            CUX1/Protein CASP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            CUX1/Protein CASP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            CUX1/Protein CASP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            CUX1/Protein CASP Antibody in Western Blot (WB)
            CUX1/Protein CASP Antibody in Western Blot (WB)
            CUX1/Protein CASP Antibody in Western Blot (WB)
            CUX1/Protein CASP Antibody in Western Blot (WB)
            CUX1/Protein CASP Antibody in Western Blot (WB)
            CUX1/Protein CASP Antibody in Western Blot (WB)
            CUX1/Protein CASP Antibody in Immunoprecipitation (IP)

            Product Details

            11733-1-AP

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:1,000-1:4,000
            View 11 publications 11 publications

            Immunohistochemistry (IHC)

            -
            View 16 publications 16 publications

            Immunohistochemistry (Paraffin) (IHC (P))

            1:50-1:500
            -

            Immunocytochemistry (ICC/IF)

            1:10-1:100
            View 18 publications 18 publications

            Immunoprecipitation (IP)

            0.5-4.0 µg
            View 1 publication 1 publication

            Not applicable (N/A)

            -
            View 1 publication 1 publication
            Product Specifications

            Species Reactivity

            Human, Mouse, Rat

            Published species

            Human

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            CUX1 Fusion Protein Ag2335 (1-341 aa encoded by BC025422)
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.13 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS, pH 7.3, with 50% glycerol

            Contains

            0.02% sodium azide

            Storage conditions

            -20°C

            Shipping conditions

            Wet ice (domestic); Dry ice (international)

            Product Specific Information

            Immunogen sequence: MAANVGSMF QYWKRFDLQQ LQRELDATAT VLANRQDESE QSRKRLIEQS REFKKNTPED LRKQVAPLLK SFQGEIDALS KRSKEAEAAF LNVYKRLIDV PDPVPALDLG QQLQLKVQRL HDIETENQKL RETLEEYNKE FAEVKNQEVT IKALKEKIRE YEQTLKNQAE TIALEKEQKL QNDFAEKERK LQETQMSTTS KLEEAEHKVQ SLQTALEKTR TELFDLKTKY DEETTAKADE IEMIMTDLER ANQRAEVAQR EAETLREQLS SANHSLQLAS QIQKAPDVAI EVLTRSSLEV ELAAKEREIA QLVEDVQRLQ ASLTKLRENS ASQISQLEQQ LS (1-341 aa encoded by BC025422)

            Target Information

            The protein encoded by this gene is a member of the homeodomain family of DNA binding proteins. It may regulate gene expression, morphogenesis, and differentiation and it may also play a role in the cell cycle progession. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: CCAAT displacement protein; CDP; CDP/Cux p200; CDP2; cut homolog; cut-like 1; cut-like homeobox 1; Cux/CDP homeoprotein; golgi integral membrane protein 6; Homeobox protein cut-like 1; Homeobox protein cux-1; Protein CASP; putative protein product of Nbla10317

            View more View less

            Gene Aliases: CASP; CDP; CDP/Cut; CDP1; Clox; COY1; CUTL1; CUX; Cux-1; Cux/CDP; CUX1; GOLIM6; Kiaa4047; Nbla10317; p100; p110; p200; p75

            View more View less

            UniProt ID: (Human) P39880, (Mouse) P53564

            View more View less

            Entrez Gene ID: (Human) 1523, (Rat) 116639, (Mouse) 13047

            View more View less

            Function(s)

            RNA polymerase II regulatory region sequence-specific DNA binding protein tyrosine kinase activity protein binding, bridging sequence-specific DNA binding transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding DNA binding chromatin binding homeodomain transcription factor

            Process(es)

            negative regulation of transcription from RNA polymerase II promoter retrograde transport, vesicle recycling within Golgi transcription, DNA-templated regulation of transcription from RNA polymerase II promoter multicellular organism development peptidyl-tyrosine phosphorylation positive regulation of dendrite morphogenesis kidney development regulation of transcription, DNA-templated transcription from RNA polymerase II promoter intra-Golgi vesicle-mediated transport lung development auditory receptor cell differentiation positive regulation of transcription from RNA polymerase II promoter transport
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-7c4bf66445-xskn5:80/100.66.75.180:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0