Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%).
This antibody has specificity for Rat CXCL1.
CXCL1, CXCL2, and CXCL3, also known respectively as GRO alpha, GRO beta (MIP-2 alpha) and GRO gamma (MIP-2 beta), are members of the CXC subfamily of chemokines. Mature CXCL1/2/3 proteins bind with high affinity to the IL-8 receptor type B and are potent neutrophil attractants and activators. In vitro, the processed forms GRO-alpha(4-73), GRO-alpha(5-73) and GRO-alpha(6-73) show a 30-fold higher chemotactic activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C Cmotif chemokine; C X C motif chemokine; C-X-C motif chemokine 1; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 1 (melanoma growth stimulating activity, alpha); CINC-1; CXC; CXC motif chemokine; CXCL; CXCL-1; Cytokine-induced neutrophil chemoattractant 1; gro; GRO a; GRO α; GROalpha; Growth-regulated alpha protein; GROα; NAP3; Platelet-derived growth factor-inducible protein KC
Gene Aliases: CINC-1; Cinc1; Cxcl1; Gro; Gro1; Scyb1
UniProt ID: (Rat) P14095
Entrez Gene ID: (Rat) 81503
Molecular Function: chemokine
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support