Invitrogen antibodies—maximize lab budgets with advanced validation & coverage

Shop antibodies

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • CXCL13 Antibodies

            Bioss

            CXCL13/BCA1 Polyclonal Antibody

            View all (24) CXCL13 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Catalog # BS-4509R
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            455.00
            100 µL
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite CXCL13/BCA1 Polyclonal Antibody

            • Antibody Testing Data (1)
            Application
            CXCL13/BCA1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Group 53 Created with Sketch.

            FIGURE: 1 / 1

            CXCL13/BCA1 Antibody (BS-4509R) in IHC (P)

            Formalin-fixed and paraffin embedded rat liver tissue labeled with Anti-CXCL13 Polyclonal Antibody, Unconjugated (bs-4509R) at 1:200 followed by conjugation to the secondary antibody and DAB staining.
            View Product

            CXCL13/BCA1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))

            Product Details

            BS-4509R

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:300-1:1,000
            -

            Immunohistochemistry (Paraffin) (IHC (P))

            1:200
            -

            Immunohistochemistry (Frozen) (IHC (F))

            1:100-1:500
            -

            Immunocytochemistry (ICC/IF)

            1:50-1:200
            -

            ELISA (ELISA)

            1:500-1:1,000
            -
            Product Specifications

            Species Reactivity

            Mouse, Rat

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            KLH conjugated synthetic peptide derived from mouse CXCL13/BCA1, amino acids 23-109.
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            1 mg/mL

            Purification

            Protein A

            Storage buffer

            0.01M TBS, pH 7.4, with 50% glycerol, 1% BSA

            Contains

            0.02% ProClin 300

            Storage conditions

            -20°C

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            Target Information

            CXCL13, also known as BCA-1 and Angie, is a small chemokine strongly expressed in the secondary lymphoid organs. It preferentially promotes the migration of B lymphocytes (both B-1 and B-2) to the follicles of the spleen, lymph nodes, and Peyer's patches. It is produced by macrophages, and in mouse the highest constitutive producers include large peritoneal macrophages, Kuppfer cells and lung macrophages (both CD11c+ and CD11c-). The cellular ligand for CXCL13 is CXCR5 also known as BLR-1 (Burkitt’s Lymphoma Receptor 1). CXCR5 is expressed on B cells, Tfh cells and, to a lower degree, some memory T cell populations. CXCL13 plays an important role in the development of chronic lymphocytic leukemia (CCL). CXCL13 is also a plasma biomarker for germinal center activity and generation of broadly neutralizing antibodies against HIV.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: B lymphocyte chemoattractant; C Cmotif chemokine; C X C motif chemokine; C-X-C motif chemokine 13; CC motif chemokine; CCmotif chemokine; chem; CXC; CXC chemokine BLC; CXC motif chemokine; CXC motif chemokine 13; CXCL; small inducible cytokine subfamily B (Cys-X-Cys), member 13; Small-inducible cytokine B13

            View more View less

            Gene Aliases: 4631412M08Rik; Angie; ANGIE2; BCA-1; BLC; BLR1L; Cxcl13; Scyb13

            View more View less

            UniProt ID: (Mouse) O55038

            View more View less

            Entrez Gene ID: (Mouse) 55985, (Rat) 498335

            View more View less

            Function(s)

            cytokine activity chemokine activity heparin binding fibroblast growth factor binding CXCR5 chemokine receptor binding CCR10 chemokine receptor binding protein heterodimerization activity receptor agonist activity CXCR3 chemokine receptor binding CXCR chemokine receptor binding chemokine

            Process(es)

            lymphocyte chemotaxis across high endothelial venule positive regulation of leukocyte chemotaxis chemotaxis inflammatory response immune response cell surface receptor signaling pathway G-protein coupled receptor signaling pathway positive regulation of cytosolic calcium ion concentration cell-cell signaling positive regulation of T cell chemotaxis response to lipopolysaccharide positive regulation of integrin activation positive regulation of cell-cell adhesion mediated by integrin B cell chemotaxis endothelial cell chemotaxis to fibroblast growth factor B cell chemotaxis across high endothelial venule regulation of cell proliferation defense response to bacterium lymph node development chemokine-mediated signaling pathway activation of GTPase activity positive regulation of receptor activity negative regulation of endothelial cell chemotaxis to fibroblast growth factor
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            United States flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-d5cbd:80/100.66.74.37:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0