Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MARATLSAAP SNPRLLRVAL LLLLLVAASR RAAGAPLATE LRCQCLQTLQ GIHLKNIQSV KVKSPGPHCA QTEVIATLKN GQKACLNPAS PMVKKIIEKM
Produced by activated monocytes and neutrophils and expressed at sites of inflammation. Hematoregulatory chemokine, which, in vitro, suppresses hematopoietic progenitor cell proliferation. GRO-beta(5-73) shows a highly enhanced hematopoietic activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C Cmotif chemokine; C X C motif chemokine; C-X-C motif chemokine 2; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 2; CXC; CXC motif chemokine; CXCL; GRO b; gro beta; GRO β; Gro-beta; GRO2 oncogene; GRObeta; Growth-regulated protein beta; GROβ; Macrophage inflammatory protein 2-alpha; melanoma growth stimulatory activity beta; MGSA beta; MIP 2 a; MIP 2 alpha; MIP 2 α; MIP2 a; MIP2 α; MIP2-alpha
Gene Aliases: CINC-2a; CXCL2; GRO2; GROB; MGSA-b; MIP-2a; MIP2; MIP2A; SCYB2
UniProt ID: (Human) P19875
Entrez Gene ID: (Human) 2920
Molecular Function: chemokine
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support