Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Antibody detects endogenous levels of total GCP2.
CXCL6 is a chemotactic for neutrophil granulocytes. It signals through binding and activation of its receptors (CXCR1 and CXCR2). In addition to its chemotactic and angiogenic properties, it has strong antibacterial activity against Gram-positive and Gram-negative bacteria (90-fold-higher when compared to CXCL5 and CXCL7).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C Cmotif chemokine; C X C motif chemokine; C-X-C motif chemokine 5; C-X-C motif chemokine 6; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 5; chemokine (C-X-C motif) ligand 6 (granulocyte chemotactic protein 2); Chemokine alpha 3; CKA-3; CKA3; CXC; CXC chemokine LIX; CXC motif chemokine; CXCL; Cytokine LIX; GCP-2; Granulocyte chemotactic protein 2; small inducible cytokine subfamily B (Cys-X-Cys), member 6 (granulocyte chemotactic protein 2); Small inducible cytokine subfamily B (Cys-X-Cys), member b; Small-inducible cytokine B5; Small-inducible cytokine B6
Gene Aliases: CKA-3; Cxcl5; CXCL6; GCP-2; GCP2; Scyb5; SCYB6
UniProt ID: (Human) P80162, (Rat) P97885
Entrez Gene ID: (Human) 6372, (Rat) 60665
Molecular Function: chemokine
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support