Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Alomone Labs, LTD
Reconstitution: 1 X 50 µL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
CXCR2 is a receptor for interleukin-8, which is a powerful neutrophil chemotactic factor. It is a member of the GPCR family. Binding of IL-8 to the receptor causes activation of neutrophils. This response is mediated via a G-protein that activate a phosphatidylinositol-calcium second messenger system. This receptor binds to IL-8 with a high affinity and to GRO/MGSA and NAP-2 also with a high affinity. It has been reported to be expressed in a wide variety of tissues. ESTs have been isolated from human placenta and thymus libraries.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C Cmotif chemokine; C X C motif chemokine; C-X-C chemokine receptor type 2; CC motif chemokine; CCmotif chemokine; CD182; CD182 antigen; CDw128b; chemokine (C-X-C motif) receptor 2; chemokine (CXC) receptor 2; CXC; CXC motif chemokine; CXC-R2; CXCR-2; CXCR2 gene for IL8 receptor type B; GRO/MGSA receptor; High affinity interleukin-8 receptor B; Il-8 receptor b; IL-8 receptor beta; IL-8 receptor type 2; IL-8R B; interleukin 8 receptor B; interleukin 8 receptor type 2; interleukin 8 receptor, beta; interleukin-8 receptor type B
Gene Aliases: CD182; CDw128b; CMKAR2; CXCR2; IL8R2; IL8RA; IL8RB
UniProt ID: (Human) P25025
Entrez Gene ID: (Human) 3579
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support