Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MMGGESADKA TAAAAAASLL ANGHDLAAAM AVDKSNPTSK HKSGAVASLL SKAERATELA AEGQLTLQQF AQSTEMLKRV VQEHLPLMSE AGAGLPDMEA VAGAEALNGQ SDFPYLGAFP INPGLFIMTP AGVFLAESAL HMAGLAEYPM QGELASAISS GKKKRKRCGM CAPCRRRINC EQCSSCRNRK TGHQICKFRK CEELKKKPSA ALEKVMLPTG AAFRWFQ
CXXC5 is zinc finger protein closely related to the DVL1-binding protein CXXC4 (also known as Idax), and like CXXC4 is thought to modulate the Wnt signaling pathway during development. CXXC5 is upregulated in neural stem cells upon BMP4 stimulation, leading to decreased levels of the Wnt-signaling target AXIN2 and facilitated the response of the stem cells to Wnt3a. Recent reports suggest CXXC5 transcription is activated by the Wilms tumor 1 (WT1) transcription factor, and together are involved in the WNT/beta-catenin pathway and play a role in embryonic kidney development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CF5; CXXC finger 5 protein; CXXC-type zinc finger protein 5; Putative MAPK-activating protein PM08; Putative NF-kappa-B-activating protein 102; Retinoid-inducible nuclear factor; RINF; WT1-induced Inhibitor of Dishevelled
Gene Aliases: CF5; CXXC5; HSPC195; RINF; TCCCIA00297; WID
UniProt ID: (Human) Q7LFL8
Entrez Gene ID: (Human) 51523
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support