Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
FIGURE: 1 / 4
CYB561D1 gene ontology (GO) annotations include oxidation-reduction process.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cytochrome b-561 domain containing 1; Cytochrome b561 domain-containing protein 1; cytochrome b561 family, member D1; Probable transmembrane reductase CYB561D1
Gene Aliases: CYB561D1
UniProt ID: (Human) Q8N8Q1
Entrez Gene ID: (Human) 284613
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support