Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Immunogen sequence: TLLDPNEKYL LRLLDKTTVS HNTKRFRFAL PTAHHTLGLP VGKHIYLSTR IDGSLVIRPY TPVTSDEDQG YVDLVIKVYL KGVHPKFPEG GKMSQYLDSL KVGDVVEFRG PSGLLTYTGK GHFNIQPNKK SPPEP
Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.
NADH-cytochrome b5 reductases are involved in desaturation and elongation of fatty acids, cholesterol biosynthesis, drug metabolism, and, in erythrocyte, methemoglobin reduction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: b5R.1; Humb5R2; NAD(P)H:quinone oxidoreductase type 3 polypeptide A2; NAD(P)H:quinone oxidoreductase type 3, polypeptide A2; NADH-cytochrome b5 reductase 1
Gene Aliases: B5R.1; B5R1; B5R2; CYB5R1; humb5R2; NQO3A2; UNQ3049/PRO9865
UniProt ID: (Human) Q9UHQ9
Entrez Gene ID: (Human) 51706
Molecular Function: reductase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support