Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: GLTEKLIFSLR DPAYSTFPPE GVF
Highest antigen sequence identity to the following orthologs - mouse 67%, rat 67%.
This gene is a member of the cytochrome b(561) family that encodes an iron-regulated protein. It highly expressed in the duodenal brush border membrane. It has ferric reductase activity and is believed to play a physiological role in dietary iron absorption.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytochrome b reductase 1; cytochrome b561 family, member A2; Duodenal cytochrome b; Ferric-chelate reductase 3; Plasma membrane ascorbate-dependent reductase CYBRD1
Gene Aliases: CYB561A2; CYBRD1; DCYTB; FRRS3
UniProt ID: (Human) Q53TN4
Entrez Gene ID: (Human) 79901
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support