Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 5
Recommended positive controls: HeLa, mouse brain, rat brain.
Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (93%), Bovine (100%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Component of the CYFIP1-EIF4E-FMR1 complex which binds to the mRNA cap and mediates translational repression. In the CYFIP1-EIF4E-FMR1 complex this subunit is an adapter between EIF4E and FMR1. Promotes the translation repression activity of FMR1 in brain probably by mediating its association with EIF4E and mRNA. Involved in formation of membrane ruffles and lamellipodia protrusions and in axon outgrowth. Binds to F-actin but not to RNA. Regulator of epithelial morphogenesis. May act as an invasion suppressor in cancers.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CYFIP1; Cytoplasmic FMR1-interacting protein 1; cytoplasmic FMRP interacting protein 1; FLJ45151; p140sra-1; selective hybridizing clone; Specifically Rac1-associated protein 1; Sra-1
Gene Aliases: CYFIP1; E030028J09Rik; KIAA0068; l(7)1Rl; l71Rl; l7Rl1; mKIAA0068; P140SRA-1; P140sra1; pl-1; SHYC; SRA-1; SRA1
UniProt ID: (Human) Q7L576, (Mouse) Q7TMB8
Entrez Gene ID: (Human) 23191, (Mouse) 20430, (Rat) 308666
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support