Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MEKVPGEMEI ERRERSEELS EAERKAVQAM WARLYASCED VGVAILVRFF VNFPSAKQYF SQFKHMEDPL EMERSPQLRK HACRVMGALN TVVENLHDPD KVSSVLALVG KAHALKHKVE PVYFKILSGV ILEVVAEEFA SDFPPETQRA WAKLRGLIYS HVTAAYKEVG WVQQVPNATT PPATLPSSGP
Cytoglobin is a ubiquitously expressed hexacoordinate hemoglobin that may facilitate diffusion of oxygen through tissues, scavenge nitric oxide or other reactive oxygen species, or serve a protective function during oxidative stress (Trent and Hargrove, 2002).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytoglobin; HGb; Histoglobin; Nitric oxygen dioxygenase CYGB; Nitrite reductase CYGB; NOD; Pseudoperoxidase CYGB; Stellate cell activation-associated protein; Superoxide dismutase CYGB
Gene Aliases: CYGB; HGB; STAP
UniProt ID: (Human) Q8WWM9
Entrez Gene ID: (Human) 114757
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support