Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MLAKGLPPRS VLVKGCQTFL SAPREGLGRL RVPTGEGAGI STRSPRPFNE IPSPGDNGWL NLYHFWRETG THKVHLHHVQ NFQKYGPIYR EKLGNVESVY VIDPEDVALL FKSEGPNPER FLIPPWVAYH QYYQRPIGVL LKKSAAWKKD RVALNQEVMA PEATKNFLPL LDAVSRDFVS VLHRRIKKAG SGNYSGDISD DLFRFAFESI TNVIFGERQG MLEEVVNPEA QRFIDAIYQM FHTSVPMLNL PPDLFRLFRT KTWKDHVAAW DVIFSKADIY TQNFYWELRQ KGSVHHDYRG ILYRLLGDSK MSFEDIKANV TEMLAGGVDT TSMTLQWHLY EMARNLKVQD MLRAEVLAAR HQAQGDMATM LQLVPLLKAS IKETLRLHPI SVTLQRYLVN DLVLRDYMIP AKTLVQVAIY ALGREPTFFF DPENFDPTRW LSKDKNITYF RNLGFGWGVR QCLGRRIAEL EMTIFLINML ENFRVEIQHL SDVGTTFNLI LMPEKPISFT FWPFNQEATQ Q
CYP11A1, also known as cytochrome P450C11A1, cytochrome P450scc and cytochrome P450, subfamily XIA, is an enzyme that catalyzes the first step of steroid biosynthesis under the modulation of cAMP signal. CYP11A1 in steroidogenic cells converts cholesterol to pregnenolone, which is determined by hormonal control of cholesterol availability. Expression of the CYP11A1 gene is controlled by the transcription factor SF-1, and the upstream SF-1 binding site in the CYP11A1 gene is required for hormonal stimulation. c-Jun and SF-1 may act synergistically to activate CYP11A1 gene expression. Both Forskolin and 8-Br-cAMPS elevate CYP11A1 mRNA levels in the interstitial cell monolayer, which has a fully functional adenylate cyclase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cholesterol 20-22 desmolase; Cholesterol desmolase; cholesterol monooxygenase (side-chain cleaving); Cholesterol side-chain cleavage enzyme, mitochondrial; CYPXIA1; Cytochrome P450 11A1; cytochrome P450 family 11 subfamily A polypeptide 1; Cytochrome P450(scc); cytochrome P450, family 11, subfamily A, polypeptide 1; cytochrome P450, subfamily XIA (cholesterol side chain cleavage); cytochrome P450C11A1; steroid 20-22-lyase
Gene Aliases: CYP11A; CYP11A1; CYPXIA1; P450SCC
UniProt ID: (Human) P05108
Entrez Gene ID: (Human) 1583
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support