Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MLFPISMSAT EFLLASVIFC LVFWVIRASR PQVPKGLKNP PGPWGWPLIG HMLTLGKNPH LALSRMSQQY GDVLQIRIGS TPVVVLSGLD TIRQALVRQG DDFKGRPDLY TFTLISNGQS MSFSPDSGPV WAARRRLAQN GLKSFSIASD PASSTSCYLE EHVSKEAEVL ISTLQELMAG PGHFNPYRYV VVSVTNVICA ICFGRRYDHN HQELLSLVNL NNNFGEVVGS GNPADFIPIL RYLPNPSLNA FKDLNEKFYS FMQKMVKEHY KTFEKGHIRD ITDSLIEHCQ EKQLDENANV QLSDEKIINI VLDLFGAGFD TVTTAISWSL MYLVMNPRVQ RKIQEELDTV IGRSRRPRLS DRSHLPYMEA FILETFRHSS FVPFTIPHST TRDTSLKGFY IPKGRCVFVN QWQINHDQKL WVNPSEFLPE RFLTPDGAID KVLSEKVIIF GMGKRKCIGE TIARWEVFLF LAILLQRVEF SVPLGVKVDM TPIYGLTMKH ACCEHFQMQL RS
This gene, CYP1A1, encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and its expression is induced by some polycyclic aromatic hydrocarbons (PAHs), some of which are found in cigarette smoke. The enzyme's endogenous substrate is unknown; however, it is able to metabolize some PAHs to carcinogenic intermediates. The gene has been associated with lung cancer risk. A related family member, CYP1A2, is located approximately 25 kb away from CYP1A1 on chromosome 15.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: aryl hydrocarbon hydroxylase; CYP; CYPIA1; cytochrome P1-450, dioxin-inducible; Cytochrome P450 1A1; Cytochrome P450 form 6; cytochrome P450, family 1, subfamily A, polypeptide 1; cytochrome P450, subfamily I (aromatic compound-inducible), polypeptide 1; Cytochrome P450-C; Cytochrome P450-P1; flavoprotein-linked monooxygenase; Hydroperoxy icosatetraenoate dehydratase; P450; P450 form 6; P450-P1; xenobiotic monooxygenase
Gene Aliases: AHH; AHRR; CP11; CYP1; CYP1A1; CYPIA1; P1-450; P450-C; P450DX
UniProt ID: (Human) P04798
Entrez Gene ID: (Human) 1543
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support