Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CYP2R1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This enzyme is a microsomal vitamin D hydroxylase that converts vitamin D into the active ligand for the vitamin D receptor. A mutation in CYP2R1 gene has been associated with selective 25-hydroxyvitamin D deficiency.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytochrome P450 2R1; cytochrome P450, 2r1; cytochrome P450, family 2, R1; cytochrome P450, family 2, subfamily R, polypeptide 1; Vitamin D 25-hydroxylase
Gene Aliases: CYP2R1
UniProt ID: (Human) Q6VVX0, (Mouse) Q6VVW9
Entrez Gene ID: (Human) 120227, (Mouse) 244209, (Rat) 361631
Molecular Function:
oxidoreductase
oxygenase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support