Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EPVADKILQ ELKCLSGQLD GYRGRPFPLA LLGWAPSNIT FALLFGRRFD YRDPVFVSLL GLIDEVMVLL GSPGLQLFNV YPWLGALLQL HRPVLRKIEE VRAILRTLLE ARRPHVCPGD PVCSYVDALI QQGQGDDPEG LFAEANAVAC TLDMVMAGTE TTSATLQWAA LLMGRHPDVQ GRVQEELDRV LGPGRTPRLE DQQALPYTSA VLHEVQRFIT LLPHVPRCTA ADTQLGGFLL PKGTPVIPLL TSVLLDETQW QTPGQFNPGH FLDANGHFVK REAFLPFSAG RRVCVGERLA RTELFLLFAG LLQRYRLLPP PGVSPASLDT TPARAFTMRP RAQALCAVPR P
CYP2W1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CP2W1; CYPIIW1; Cytochrome P450 2W1; cytochrome P450, family 2, subfamily W, polypeptide 1
Gene Aliases: CYP2W1; Gm455
UniProt ID: (Human) Q8TAV3, (Mouse) E9Q816
Entrez Gene ID: (Human) 54905, (Mouse) 545817, (Rat) 288517
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support