Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: YGDVLVRNL RREAEKGKPV TLKDIFGAYS MDVITGTSFG VNIDSLNNPQ DPFVESTKKF LKFGFLDPLF LSIILFPFLT PVFEALNVSL FPKDTINFLS KSVNRMKKSR LNDKQKHRLD FLQLMIDSQN SKETESHKAL SDLELAAQSI IFIFAGYETT SSVLSFTLYE LATHPDVQQK LQKEIDAVLP NKAPPTYDAV VQMEYLDMVV NETLRLFPVA IRLERTCKKD VEINGVFIPK GSMVVIPTYA LHHDPKYWTE PEEFRPERFS KKKDSIDPYI YTPFGTGPRN CIGMRFALMN MKLALIRVLQ NFSFKPCKET QIPLKLDTQG LLQPEKPIVL KVDSRDGTLS GE (152-502 aa encoded by BC033862)
CYP3A5 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and its expression is induced by glucocorticoids and some pharmacological agents. The enzyme metabolizes drugs such as nifedipine and cyclosporine as well as the steroid hormones testosterone, progesterone and androstenedione.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: aryl hydrocarbon hydroxylase; CYPIIIA5; Cytochrome P450 3A5; cytochrome P450 HLp2; cytochrome P450, family 3, subfamily A, polypeptide 5; cytochrome P450, subfamily IIIA (niphedipine oxidase), polypeptide 5; Cytochrome P450-PCN3; flavoprotein-linked monooxygenase; HLp2; microsomal monooxygenase; P450-PCN3; xenobiotic monooxygenase
Gene Aliases: CP35; CYP3A5; CYPIIIA5; P450PCN3; PCN3
UniProt ID: (Human) P20815
Entrez Gene ID: (Human) 1577
Molecular Function:
oxygenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support