Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Cytochrome P450 super family proteins are monooxygenases which catalyze multiple biochemical reactions involved in drug metabolism and synthesis of cholesterol, steroids and lipids. CYP4F3, a 60 kDa protein, is a novel member of this family. It consists of a conserved heme-binding domain near the C-terminus and catalyzes the u-hydroxylation of LTB4. CYP4F3 is localized in the ER and plays a vital role in the inactivation of LTB4 and controlling inflammation. CYP4F3 exists in two functionally distinct isoforms: CYP4F3A, a neutrophil isoform, is particularly expressed in neutrophils and myeloid cells and catalyzes the microsomal u-hydroxylation of LTB4. It interacts with CD11b and myeloperoxidase on myeloid cells and regulates cellular development in bone marrow. CYP4F3B, a liver isoform, catalyzes the conversion of arachidonic acid to 20-HETE, a potent activator of PKC. Its role has been implicated in cell proliferation, vascular tone and natriuresis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 20-HETE synthase; 20-hydroxyeicosatetraenoic acid synthase; CYPIVF3; cytochrome P-450; Cytochrome P450 4F3; cytochrome P450, family 4, subfamily F, polypeptide 3; cytochrome P450, subfamily IVF, polypeptide 3 (leukotriene B4 omega hydroxylase); Cytochrome P450-LTB-omega; Docosahexaenoic acid omega-hydroxylase CYP4F3; leukotriene B4 omega hydroxylase; Leukotriene-B(4) 20-monooxygenase 2; Leukotriene-B(4) omega-hydroxylase 2; leukotriene-B4 20-monooxygenase
Gene Aliases: CPF3; CYP4F; CYP4F3; LTB4H
UniProt ID: (Human) Q08477
Entrez Gene ID: (Human) 4051
Molecular Function: oxygenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support