Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CYP4F8 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and functions as a 19-hydroxylase of prostaglandins in seminal vesicles.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CYPIVF8; Cytochrome P450 4F8; cytochrome P450, family 4, subfamily F, polypeptide 8; cytochrome P450, subfamily IVF, polypeptide 8; flavoprotein-linked monooxygenase; microsomal monooxygenase
Gene Aliases: CPF8; CYP4F8; CYPIVF8
UniProt ID: (Human) P98187
Entrez Gene ID: (Human) 11283
Molecular Function:
oxygenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support