Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The cysteinyl leukotrienes LTC4, LTD4, and LTE4 are important mediators of human bronchial asthma. Pharmacologic studies have determined that cysteinyl leukotrienes activate at least 2 receptors, the protein encoded by this gene and CYSLTR2. This encoded receptor is a member of the superfamily of G protein-coupled receptors. Activation of this receptor by LTD4 results in contraction and proliferation of smooth muscle, oedema, eosinophil migration and damage to the mucus layer in the lung.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cys-LT1 receptor; CysLT 1; CysLT(1); CysLT1 receptor; CysLTR1; cysteinyl leukotriene 1 receptor short isoform; Cysteinyl leukotriene D4 receptor; Cysteinyl leukotriene receptor 1; G-protein coupled receptor HG55; HG55; HMTMF81; LTD4 receptor; LTD4R; MGC46139
Gene Aliases: BB147369; CYSLT1; CYSLT1R; CYSLTR; CYSLTR1; HMTMF81
UniProt ID: (Human) Q9Y271, (Mouse) Q99JA4
Entrez Gene ID: (Human) 10800, (Mouse) 58861
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support