Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Recommended positive controls: U87-MG, SK-N-SH, IMR32, SK-N-AS, 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji, Mouse brain, Rat brain.
Predicted reactivity: Mouse (98%), Rat (98%), Pig (96%), Bovine (99%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CAMK1G serine/threonine kinase is a member of CAMK family and is a novel membrane-anchored neuronal Ca2+ calmodulin-dependent protein kinase. Full activation of CAMK1G requires both Ca2+ and calmodulin and phosphorylation by CAMKK. CAMK1G transcripts are most abundant in neurons, with the highest levels in limited nuclei such as the central nucleus of the amygdala (CeA) and the ventromedial hypothalamus.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calcium/calmodulin-dependent protein kinase I gamma; calcium/calmodulin-dependent protein kinase IG; Calcium/calmodulin-dependent protein kinase type 1G; CaM kinase 1G; CaM kinase I gamma; CaM kinase IG; caM-KI gamma; CaMK-like CREB kinase III; caMKI gamma; caMKI-gamma; caMKIG; CLICK III; RP1-272L16.2
Gene Aliases: CAMK1G; Camkig; CaMKIgamma; CLICK-III; CLICK3; CLICKIII; dJ272L16.1; VWS1
UniProt ID: (Human) Q96NX5, (Mouse) Q91VB2, (Rat) Q7TNJ7
Entrez Gene ID: (Human) 57172, (Mouse) 215303, (Rat) 171358
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support