Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The expression of multiple classes of voltage dependent calcium channels (VDCCs) allows neurons to tailor calcium signaling to functionally discrete cellular regions. Although N-Type VDCCs exist before birth which is consistent with a role in migration, most N-Type VDCCs subunit expression is postnatal and is important in the genesis of synaptic transmission in discrete hippocampal fields.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BII; Brain calcium channel II; Calcium channel, L type, alpha-1 polypeptide, isoform 6; calcium channel, R type, alpha-1 polypeptide, isoform 6; voltage-dependent calcium channel alpha 1E subunit; Voltage-dependent R-type calcium channel subunit alpha-1E; voltage-gated calcium channel alpha subunit Cav2.3; Voltage-gated calcium channel subunit alpha Cav2.3
Gene Aliases: BII; CACH6; CACNA1E; CACNL1A6; Cav2.3; gm139
UniProt ID: (Human) Q15878
Entrez Gene ID: (Human) 777
Molecular Function:
voltage-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support