Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 4
Immunogen sequence: MDAVDATME KLRAQCLSRG ASGIQGLARF FRQLDRDGSR SLDADEFRQG LAKLGLVLDQ AEAEGVCRKW DRNGSGTLDL EEFLRALRPP MSQAREAVIA AAFAKLDRSG DGVVTVDDLR GVYSGRAHPK VRSGEWTEDE VLRRFLDNFD SSEKDGQVTL AEFQDYYSGV SASMNTDEEF VAMMTSAWQL (1-189 aa encoded by BC080599)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Calcyphosin; Calcyphosine; calcyphosine 1; thyroid protein p24
Gene Aliases: CAPS; CAPS1
UniProt ID: (Human) Q13938
Entrez Gene ID: (Human) 828
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support