Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: FWLPLLEKVY AKVHGSYEHL WAGQVADALV DLTGGLAERW NLKGVAGSGG QQDRPGRWEH RTCRQLLHLK DQCLISCCVL SPRAGARELG EFHAFIVSDL
Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 82%.
The calpains including Calpain 1 (CAPN1), calcium-activated neutral proteases, are nonlysosomal, intracellular cysteine proteases. The mammalian calpains include ubiquitous, stomach-specific, and muscle-specific proteins. The ubiquitous enzymes consist of heterodimers with distinct large, catalytic subunits associated with a common small, regulatory subunit. This gene encodes the large subunit of the ubiquitous enzyme, calpain 1. Several transcript variants encoding two different isoforms have been found for this gene. Diseases associated with CAPN1 include Spastic Paraplegia 76, Autosomal Recessive and Spasticity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Calcium-activated neutral proteinase 10; Calpain-10; calpain-like protease CAPN10; CANP 10
Gene Aliases: CANP10; CAPN10; KIAA1845; NIDDM1
UniProt ID: (Human) Q9HC96
Entrez Gene ID: (Human) 11132
Molecular Function:
cysteine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support