Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEICADELK KVLNTVVNKH KDLKTHGFTL ESCRSMIALM DTDGSGKLNL QEFHHLWNKI KAWQKIFKHY DTDQSGTINS YEMRNAVNDA GFHLNNQLYD IITMRYADKH MNIDFDSFIC CFVRLEGMFR AFHAFDKDGD GIIKLNVLEW LQLTMYA (1-156 aa encoded by BC004883)
Calpain, an intracellular calcium-dependent protease that cleaves cytoskeletal and submembranous proteins, plays a role in cytoskeletal reorganization and muscle protein degradation. Calpain is a heterodimer composed of a small regulatory subunit and one of three large catalytic subunits, designated calpain 1, calpain 2 and calpain 3. Calpain 3 (calpain p94) is a muscle-preferred calcium activated neutral protease that localizes to the nucleus. Mutations involving the calpain 3 gene are associated with limb-girdle muscle dystrophy type 2A, a form of autosomal recessive and progressive neuromuscular diso4. Calpastatin regulates calpain by inhibiting both the proteolytic activity of calpain and its binding to membranes. Calpastatin exists in two types, tissue type and erythrocyte type, resulting from both alternative splicing and proteolytic processing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Calcium-activated neutral proteinase 3; calpain 3, (p94); Calpain L3; Calpain p94; calpain p94, large [catalytic] subunit; calpain, large polypeptide L3; Calpain-3; CANP 3; CAPN 3; LGMD 2A; MGC10767; MGC11121; MGC14344; MGC4403; Muscle-specific calcium-activated neutral protease 3; muscle-specific calcium-activated neutral protease 3 large subunit; nCL-1; New calpain 1; skeletal muscle specific calpain p94
Gene Aliases: AI323605; CANP3; CANPL3; Capa-3; Capa3; CAPN3; LGMD2; LGMD2A; Lp82; Lp84; Lp85; nCL-1; NCL1; p94
UniProt ID: (Human) P20807, (Mouse) Q64691, (Rat) P16259
Entrez Gene ID: (Human) 825, (Mouse) 12335, (Rat) 29155
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support