Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 2
This antibody specifically recognizes the 28 to 30 kD small subunit of Calpain 1 and Calpain 2 (mu calpain and m calpain). It recognizes both native and denatured protein in ELISA, Western Blot and Immunoprecipitation.
The calpains are calcium-dependent cysteine proteases that are widely expressed in mammalian systems. Both m-calpain (calpain II) and m-calpain (calpain I) are composed of an 80 kD subunit and a 30 kD subunit. Whereas the 30 kDa subunit is shared by both enzymes, the larger catalytic subunits are different and exhibit the distinct Ca++ requirements that are suggested by their names. Whereas m-calpain requires millimolar (mM) levels of calcium, m-calpain is active at micromolar (mM) concentrations of Ca++. In addition to the ubiquitously expressed m- and m- calpains, some tissue-specific calpains have been identified. The calpains appear to serve multiple physiological roles, and ideas concerning the functions of these enzymes are in a state of rapid flux.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Calcium-activated neutral proteinase small subunit; Calcium-dependent protease small subunit; Calcium-dependent protease small subunit 1; Calcium-dependent protease small subunit 2; calcium-dependent protease, small subunit; calpain 4, small subunit (30K); calpain II regulatory subunit; calpain II regulatory subunit (EC 3.4.22.17); Calpain regulatory subunit; Calpain small subunit 1; Calpain small subunit 2; calpain, small polypeptide; calpain, small subunit 1; calpain, small subunit 2; CANP small subunit; CDPS; CSS1; CSS2
Gene Aliases: CALPAIN4; CANP; CANPS; CAPN4; CAPNS; CAPNS1; CAPNS2; CDPS; CSS1
UniProt ID: (Human) P04632, (Bovine) P13135, (Human) Q96L46
Entrez Gene ID: (Human) 826, (Bovine) 281664, (Human) 84290
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support