Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 3
Calr or Calreticulin is a calcium-binding chaperone that promotes folding, oligomeric assembly and quality control in the endoplasmic reticulum (ER) via the calreticulin/calnexin cycle. Calr, a lectin, interacts transiently with almost all of the monoglucosylated glycoproteins that are synthesized in the ER. Calr also interacts with with the DNA-binding domain of NR3C1 and mediates its nuclear export. It may also be involved in maternal gene expression regulation and oocyte maturation via the regulation of calcium homeostasis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Autoantigen RO; CABP3; CALBP; Calcium-binding protein 3; Calregulin; Calreticulin; CRP55; Endoplasmic reticulum resident protein 60; epididymis secretory sperm binding protein Li 99n; ERp60; FLJ26680; grp60; HACBP; Ro/SS-A Antigen; Sicca syndrome antigen A (autoantigen Ro; calreticulin)
Gene Aliases: CALR; Calregulin; cC1qR; CRT; CRTC; HEL-S-99n; RO; SSA
UniProt ID: (Human) P27797, (Mouse) P14211, (Rat) P18418
Entrez Gene ID: (Human) 811, (Mouse) 12317, (Rat) 64202
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support