Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Calretinin is encoded by the CALB2 gene located on chromosome 16. It is a troponin C superfamily protein having six EF-hand structural motifs. The EF motif allows the protein to bind calcium, and function in calcium signaling, calcium buffering, and modulating neuronal excitability. Calretinin is expressed in the mesothelium, mast cells, some neural cells, hair follicles, and fat cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 29 kDa calbindin; 29kD calretinin; 29kDa calbindin; calbindin 2, (29kD, calretinin); calbindin 2, 29kDa; calbindin D29K; Calretinin; CR; testicular secretory protein Li 8
Gene Aliases: CAB29; CAL2; CALB2; CR
UniProt ID: (Human) P22676, (Bovine) Q3ZBY3, (Rat) P47728, (Mouse) Q08331
Entrez Gene ID: (Human) 794, (Bovine) 513947, (Horse) 100068401, (Pig) 100127479, (Rat) 117059, (Mouse) 12308
Molecular Function:
calmodulin-related
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support