Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Capricious may be involved in axon guidance, cell migration and adhesion according to gene ontology information.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Capricious,isoform A; caps; caps-PA; caps-PB; caps-PD; caps-PE; CG11282-PA; CG11282-PB; CG11282-PD; CG11282-PE; RE33792p
Gene Aliases: BcDNA:LD07388; CAPS; CG11282; CT31495; Dmel\CG11282; Dmel_CG11282; l(3)02937; l(3)rM632
Entrez Gene ID: (Fruit fly) 39493
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support