Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
Immunogen sequence: CAWQTSNNTL HPLWTVPVSV PGGTRQSPIN IQWRDSVYDP QLKPLRVSYE AASCLYIWNT GYLFQVEFDD ATEASGISGG PLENHYRLKQ FHFHWGAVNE GGSEHTVDGH AYPAELHLVH WNSVKYQNYK EAVVGENGLA VIGVFLKLGA HHQTLQRLVD ILPEIKHKDA RAAMRPFDPS TLLPTCWDYW TYAGSLTTPP LTESVTWIIQ KEPVEVAPSQ LSAFRTLLFS ALGEEEKMMV NNYRPLQPLM NRKVWASFQA TNEGTRS; Positive Samples: K-562, LO2, HepG2; Cellular Location: Mitochondrion
Carbonic anhydrases (CAs) are members of a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. CAs are involved in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric juice. They show extensive diversity in distribution and in their subcellular localization. The human CA5A gene encodes for the CA V protein which is expressed in the mitochondrial matrix in liver tissues, but can also be detected in many other tissues. In ureagenesis, the CA V protein may be involved in providing bicarbonate to carbamoyl phosphate synthetase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CA Y; CA-VA; Carbonate dehydratase VA; carbonic anhydrase 5, mitochondrial; Carbonic anhydrase 5A, mitochondrial; carbonic anhydrase 5b, mitochondrial; Carbonic anhydrase VA
Gene Aliases: Ca5; Ca5a; Car5; Car5a; CAV
UniProt ID: (Mouse) P23589
Entrez Gene ID: (Mouse) 12352
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support