Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Recommended positive controls: AsPC-1, mouse pancreas, rat pancreas.
Predicted reactivity: Dog (84%), Pig (81%), Bovine (82%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Three different procarboxypeptidases A and two different procarboxypeptidases B have been isolated. The B1 and B2 forms differ from each other mainly in isoelectric point. Carboxypeptidase B1 is a highly tissue-specific protein and is a useful serum marker for acute pancreatitis and dysfunction of pancreatic transplants. It is not elevated in pancreatic carcinoma.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Carboxypeptidase B; carboxypeptidase B1 (tissue); Pancreas-specific protein; pancreatic carboxypeptidase B; pancreatic carboxypeptidase B1; PASP; procarboxypeptidase B; protaminase; tissue carboxypeptidase B
Gene Aliases: 0910001A18Rik; 1810063F02Rik; 2210008M23Rik; AI504870; CPB; CPB1; PASP; PCPB
UniProt ID: (Human) P15086
Entrez Gene ID: (Human) 1360, (Mouse) 76703, (Rat) 24271
Molecular Function:
metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support