Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: FLAQYLCNE YQKGNETIVN LIHSTRIHIM PSLNPDGFEK AASQPGELKD WFVGRSNAQG IDLNRNFPDL DRIVYVNEKE GGPNNHLLKN MKKIVDQNTK LAPETKAVIH WIMDIPFVLS ANLHGGDLVA NYPYDETRSG SAHEYSSSPD DAIFQSLARA YSSFNPAMSD PNRPPCRKND DDSSFVDGTT NGGAWYSVPG GMQDFNYLSS NCFEITVELS CEKFPPEETL KTYWEDNKNS LISYLEQIHR GVKGFVRDLQ GNPIANATIS VEGIDHDVTS AKDGDYWRLL IPGNYKLTAS APGYLAITKK VAVPYSPAAG VDFELESFSE RKEEEKEELM EWWKMMSETL NF (126-476 aa encoded by BC033866)
This gene encodes a carboxypeptidase that cleaves C-terminal amino acid residues and is involved in the biosynthesis of peptide hormones and neurotransmitters, including insulin. It is a peripheral membrane protein. The protein specifically binds regulated secretory pathway proteins, including prohormones, but not constitutively secreted proteins. Mutations in this gene are implicated in type II diabetes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Carboxypeptidase E; Carboxypeptidase H; CarboxypeptidaseE; CarboxypeptidaseH; CBPE; cobalt-stimulated chromaffin granule carboxypeptidase; CPE; CPH; Cph 1; Enkephalin convertase; insulin granule-associated carboxypeptidase; Prohormone-processing carboxypeptidase
Gene Aliases: CARBE; CPE; CPH; Cph-1; Cph1; fat; R74677
UniProt ID: (Human) P16870, (Rat) P15087, (Mouse) Q00493
Entrez Gene ID: (Human) 1363, (Rat) 25669, (Mouse) 12876
Molecular Function: protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support