Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Immunogen sequence: GKNHTQSPGC EESDAGKEEL MKMRLPTRSD MICGYACLKG NAAMRNTKRG SWYIEALTQV FSERACDMHV ADMLVKVNAL IKEREGYAPG TEFHRCKEMS EYCSTLCQQL YLFPGYPPT
Caspases are cysteine proteases, expressed as inactive precursors, that mediate apoptosis by proteolysis of specific substrates. Caspases have the ability to cleave after aspartic acid residues. There are two classes of caspases involved in apoptosis; initiators (activation by receptor cluster) and effectors (activation by mitochondrial permeability transition). Proapoptotic signals autocatalytically activate initiator caspases, such as Caspase-8 and Caspase 9. Activated initiator caspases then process effector caspases, such as Caspase-3 and Caspase-7, which in turn cause cell collapse.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CASP-2; caspase 2 apoptosis-related cysteine peptidase; Caspase-2; ICH-1L; NEDD-2; Neural precursor cell expressed developmentally down-regulated protein 2; Protease ICH-1; protein phosphatase 1, regulatory subunit 57
Gene Aliases: CASP-2; CASP2; ICH1; NEDD-2; NEDD2; PPP1R57
UniProt ID: (Human) P42575
Entrez Gene ID: (Human) 835
Molecular Function:
protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support