Product References
Mucosal Recovery after Intestinal Transplantation in the Rat: A Sequential Histological and Molecular Assessment.
European surgical research. Europaische chirurgische Forschung. Recherches chirurgicales europeennes
Bagge J,Padma AM,Casselbrant A,Hellström M,Oltean M
43-7800 was used in Immunohistochemistry to suggest that a near-complete morphologic recovery from a moderate mucosal injury occurs within the first 24 hours after intestinal transplantation.
Mon Jun 26 00:00:00 UTC 2023
2-Methoxyestradiol combined with ascorbic acid facilitates the apoptosis of chronic myeloid leukemia cells via the microRNA-223/Fms-like tyrosine kinase 3/phosphatidylinositol-3 kinase/protein kinase B axis.
Bioengineered
Zhang S,Yu H,Li J,Fan J,Chen J
Published figure using Caspase 3 monoclonal antibody (Product # 43-7800) in Western Blot
Tue Feb 01 00:00:00 UTC 2022
Metabolic Profiling, Chemical Composition, Antioxidant Capacity, and In Vivo Hepato- and Nephroprotective Effects of Sonchus cornutus in Mice Exposed to Cisplatin.
Antioxidants (Basel, Switzerland)
Elhady SS,Abdelhameed RFA,Mehanna ET,Wahba AS,Elfaky MA,Koshak AE,Noor AO,Bogari HA,Malatani RT,Goda MS
43-7800 was used in Immunohistochemistry to investigate the potential protective effects of Sonchus cornutus extract against cisplatin-induced hepatic and renal toxicity in experimental mice.
Fri Apr 22 00:00:00 UTC 2022
Compatibility of ingredients of Danshen (Radix Salviae Miltiorrhizae) and Honghua (Flos Carthami) and their protective effects on cerebral ischemia-reperfusion injury in rats.
Experimental and therapeutic medicine
Wan H,Yang Y,Li Z,Cheng L,Ding Z,Wan H,Yang J,Zhou H
43-7800 was used in Immunohistochemistry to study the protective effects of the effective ingredients in the Chinese medicines, Danshen and Honghua in cerebrovascular disease.
Sun Aug 01 00:00:00 UTC 2021
The Ganglioside Monosialotetrahexosylganglioside Protects Auditory Hair Cells Against Neomycin-Induced Cytotoxicity Through Mitochondrial Antioxidation: An in vitro Study.
Frontiers in cellular neuroscience
Li Y,Li A,Wang C,Jin X,Zhang Y,Lu L,Wang SL,Gao X
Published figure using Caspase 3 monoclonal antibody (Product # 43-7800) in Immunocytochemistry
Fri Oct 15 00:00:00 UTC 2021
The Ganglioside Monosialotetrahexosylganglioside Protects Auditory Hair Cells Against Neomycin-Induced Cytotoxicity Through Mitochondrial Antioxidation: An in vitro Study.
Frontiers in cellular neuroscience
Li Y,Li A,Wang C,Jin X,Zhang Y,Lu L,Wang SL,Gao X
Published figure using Caspase 3 monoclonal antibody (Product # 43-7800) in Immunocytochemistry
Fri Oct 15 00:00:00 UTC 2021
Antibody-drug nanoparticle induces synergistic treatment efficacies in HER2 positive breast cancer cells.
Scientific reports
Abedin MR,Powers K,Aiardo R,Barua D,Barua S
Published figure using Caspase 3 monoclonal antibody (Product # 43-7800) in Western Blot
Thu Apr 01 00:00:00 UTC 2021
Antibody-drug nanoparticle induces synergistic treatment efficacies in HER2 positive breast cancer cells.
Scientific reports
Abedin MR,Powers K,Aiardo R,Barua D,Barua S
Published figure using Caspase 3 monoclonal antibody (Product # 43-7800) in Western Blot
Thu Apr 01 00:00:00 UTC 2021
Allium hookeri Extracts Improve Scopolamine-Induced Cognitive Impairment via Activation of the Cholinergic System and Anti-Neuroinflammation in Mice.
Nutrients
Choi JH,Lee EB,Jang HH,Cha YS,Park YS,Lee SH
43-7800 was used in Immunohistochemistry to investigate the activation of the cholinergic system and the anti-neuroinflammation effects of allium hookeri on scopolamine-induced memory impairment in mice.
Mon Aug 23 00:00:00 UTC 2021
Iron oxides nanoparticles (IOs) exposed to magnetic field promote expression of osteogenic markers in osteoblasts through integrin alpha-3 (INTa-3) activation, inhibits osteoclasts activity and exerts anti-inflammatory action.
Journal of nanobiotechnology
Marycz K,Sobierajska P,Roecken M,Kornicka-Garbowska K,Kępska M,Idczak R,Nedelec JM,Wiglusz RJ
Published figure using Caspase 3 monoclonal antibody (Product # 43-7800) in Western Blot
Tue Feb 18 00:00:00 UTC 2020
Fe3O4 Magnetic Nanoparticles Under Static Magnetic Field Improve Osteogenesis via RUNX-2 and Inhibit Osteoclastogenesis by the Induction of Apoptosis.
International journal of nanomedicine
Marycz K,Sobierajska P,Wiglusz RJ,Idczak R,Nedelec JM,Fal A,Kornicka-Garbowska K
43-7800 was used in Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot to reveal cytocompatibility and anti-inflammatory effects of fabricated nanoparticles.
Tue Oct 11 00:00:00 UTC 2022
5-Azacytydine and resveratrol reverse senescence and ageing of adipose stem cells via modulation of mitochondrial dynamics and autophagy.
Journal of cellular and molecular medicine
Kornicka K,Szłapka-Kosarzewska J,Śmieszek A,Marycz K
43-7800 was used in Western Blotting to evaluate whether treatment of ASCsEMS with a combination of 5-azacytydine and resveratrol would reverse aged phenotype of these cells.
Tue Jan 01 00:00:00 UTC 2019
Diazoxide inhibits of ER stress‑mediated apoptosis during oxygen‑glucose deprivation in vitro and cerebral ischemia‑reperfusion in vivo.
Molecular medicine reports
Lei X,Lei L,Zhang Z,Cheng Y
Published figure using Caspase 3 monoclonal antibody (Product # 43-7800) in Western Blot
Fri Jun 01 00:00:00 UTC 2018
Excessive Endoplasmic Reticulum Stress Correlates with Impaired Mitochondrial Dynamics, Mitophagy and Apoptosis, in Liver and Adipose Tissue, but Not in Muscles in EMS Horses.
International journal of molecular sciences
Marycz K,Kornicka K,Szlapka-Kosarzewska J,Weiss C
Published figure using Caspase 3 monoclonal antibody (Product # 43-7800) in Western Blot
Sat Jan 06 00:00:00 UTC 2018
Ultrastructural changes during osteogenic differentiation in mesenchymal stromal cells cultured in alginate hydrogel.
Cell & bioscience
Grzesiak J,Śmieszek A,Marycz K
437800 was used in immunocytochemistry to analyze changes occurring during the osteogenic differentiation of mesenchymal stem cells cultured in alginate hydrogel
Thu Oct 01 00:00:00 UTC 2020
Tunneling nanotubes: an alternate route for propagation of the bystander effect following oncolytic viral infection.
Molecular therapy oncolytics
Ady J,Thayanithy V,Mojica K,Wong P,Carson J,Rao P,Fong Y,Lou E
43-7800 was used in Immunohistochemistry to provide proof-of-principle demonstration of a previously unknown and alternative mechanism for inducing apoptosis in noninfected recipient cells.
Tue Sep 29 00:00:00 UTC 2020