Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 6
Immunogen sequence: ASQLERCKLG SETGSMSSMS SAEEQFQWQS QDGQKDIEDE LTTGLELVDS CIRSLQESGI LDPQDYSTGE RPSLLSQSAL QLNSKPEGSF QYPASYHSNQ TLALGETTPS QLPARGTQAR ATGQSFSQGT TSRA
Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.
Armadillo-like proteins are characterized by a series of armadillo repeats, first defined in the Drosophila 'armadillo' gene product, that are typically 42 to 45 amino acids long. These proteins can be divided into subfamilies based on the number of repeats, their overall sequence similarity, and the dispersion of the repeats throughout their sequences. Members of the p120(ctn)/plakophilin subfamily of Armadillo-like proteins, including CTNND1, Catenin delta-2, PKP1, PKP2, PKP4, and ARVCF, have 10 armadillo repeats each (Bonne et al., 1998 [PubMed 9721216]).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: catenin (cadherin-associated protein), delta 2 (neural plakophilin-related arm-repeat protein); Catenin delta-2; Delta-catenin; GT24; Neural plakophilin-related ARM-repeat protein; Neurojungin; NPRAP; T-cell delta-catenin
Gene Aliases: CTNND2; GT24; NPRAP
UniProt ID: (Human) Q9UQB3
Entrez Gene ID: (Human) 1501
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support