Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Cathepsin B is a lysosomal cysteine protease that has been implicated in the pathogenesis of rheumatoid arthritis, muscular dystrophy and tumor metastasis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: amyloid precursor protein secretase; APP secretase; APPS; Cathepsin B; Cathepsin B1; cysteine protease
Gene Aliases: APPS; CPSB; CTSB
UniProt ID: (Human) P07858
Entrez Gene ID: (Human) 1508
Molecular Function:
cysteine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support