Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: LPDSVDWREK GCVTEVKYQG SCGACWAFSA VGALEAQLKL KTGKLVSLSA QNLVDCSTEK YGNKGCNGGF MTTAFQYIID NKGIDSDASY PYKAMDQKCQ YDSKYRAATC SKYTELPYGR EDVLKEAVAN KGPVSVGVDA RHPSFFLYRS GVYYEPSCTQ NVNHGVLVVG YGDLNGKEYW LVKNSWGHNF GEEGYIRMAR NKGNHCGIAS FPSYPEI
The protein CTSS, a member of the peptidase C1 family, is a lysosomal cysteine proteinase that may participate in the degradation of antigenic proteins to peptides for presentation on MHC class II molecules. The protein can function as an elastase over a broad pH range in alveolar macrophages.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cathepsin S
Gene Aliases: CTSS
UniProt ID: (Human) P25774
Entrez Gene ID: (Human) 1520
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support