Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockout to ensure that the antibody binds to the antigen stated.
Specificity: reacts to human Caveolin-1.
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Caveolin-1 or CAV1 is a scaffolding proteins within caveolar membranes. It interacts directly with G-protein alpha subunits and can functionally regulate their activity. Caveolin-1 is also involved in the co-stimulatory signal essential for T-cell receptor (TCR) mediated T-cell activation. It can bind with DPP4 and induces T-cell proliferation and NF-kappa-B activation in a T-cell receptor/CD3-dependent manner. Mutations affecting the CAV1 gene can result in congenital generalized lipodystrophy 3, pulmonary hypertension primary 3, or partial lipodystrophy/congenital cataracts, and neurodegeneration syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: caveolae; caveolin 1, caveolae protein, 22kDa; Caveolin-1; cell growth-inhibiting protein 32; HGNC:1527; OTTHUMP00000196475
Gene Aliases: BSCL3; CAV; CAV1; CGL3; LCCNS; MSTP085; PPH3; VIP21
UniProt ID: (Human) Q03135
Entrez Gene ID: (Human) 857
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support