Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 2
This antibody is predicted to react with Monkey, Cat, Bovine.
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Caveolin-3 (CAV3) acts as a scaffolding protein within the caveolar membrane. It interacts directly with G-protein alpha subunits and can functionally regulate their activity. Caveolin-3 may also regulate voltage-gated potassium channels. It also plays a role in the sarcolemma repair mechanism of both skeletal muscle and cardiomyocytes that permits rapid resealing of membranes disrupted by mechanical stress. Caveolin-3 also mediates the recruitment of CAVIN2 and CAVIN3 proteins to the caveolae. Mutations in the gene can result in HyperCKmia, rippling muscle disease 2, cardiomyopathy, long QT syndrome 9, sudden infant death syndrome, and myopathy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAV; Caveolin-3; M-caveolin; MGC126100; MGC126101; MGC126129
Gene Aliases: AI385751; Cav-3; CAV3; LGMD1C; LQT9; M-cav; VIP-21; VIP21
UniProt ID: (Human) P56539, (Rat) P51638, (Mouse) P51637
Entrez Gene ID: (Human) 859, (Rat) 29161, (Mouse) 12391
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support