Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Immunogen sequence: TPTKKEHQKA WALNLNGFDV EEAKILRLSG KPQNAPEGYQ NRLKVLYSQK ATPGSSRKTC RYIPSLPDRI LDAPEIRNDY
Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 98%.
Required for full ubiquitin ligase activity of the anaphase promoting complex/cyclosome (APC/C) and may confer substrate specificity upon the complex. Is regulated by MAD2L1: in metaphase the MAD2L1-CDC20-APC/C ternary complex is inactive and in anaphase the CDC20-APC/C binary complex is active in degrading substrates. The CDC20-APC/C complex positively regulates the formation of synaptic vesicle clustering at active zone to the presynaptic membrane in postmitotic neurons. CDC20-APC/C-induced degradation of NEUROD2 induces presynaptic differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDC20 cell division cycle 20 homolog; cell division cycle 20 homolog; Cell division cycle protein 20 homolog; p55CDC
Gene Aliases: bA276H19.3; CDC20; CDC20A; p55CDC
UniProt ID: (Human) Q12834
Entrez Gene ID: (Human) 991
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support