Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: HeLa.
Predicted reactivity: Rhesus Monkey (100%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CDC37L1 is a cytoplasmic phosphoprotein that exists in complex with HSP90 (HSPCA; MIM 140571) as well as several other proteins involved in HSP90-mediated protein folding (Scholz et al., 2001 [PubMed 11413142]).[supplied by OMIM, Mar 2008]
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDC37 cell division cycle 37 homolog-like 1; cell division cycle 37 homolog-like 1; cell division cycle 37-like 1; Hsp90 co-chaperone Cdc37-like 1; Hsp90-associating relative of Cdc37; RP11-6J24.5
Gene Aliases: CDC37B; CDC37L1; HARC
UniProt ID: (Human) Q7L3B6
Entrez Gene ID: (Human) 55664
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support