Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MPVKRSLKL DGLLEENSFD PSKITRKKSV ITYSPTTGTC QMSLFASPTS SEEQKHRNGL SNEKRKKLNH PSLTESKEST TKDNDEFMML LSKVEKLSEE IMEIMQNLSS IQALEGSREL ENLIGISCAS HFLKREMQKT KELMTKVNKQ KLFEKSTGLP HKASRHLDSY EFLKAILN (1-177 aa encoded by BC009929)
ITGB3BP is a transcription coregulator that can have both coactivator and corepressor functions. Isoform 1, but not other isoforms, is involved in the coactivation of nuclear receptors for retinoid X (RXRs) and thyroid hormone (TRs) in a ligand-dependent fashion. ITGB3BP acts as a transcriptional corepressor via its interaction with the NFKB1 NF-kappa-B subunit, possibly by interfering with the transactivation domain of NFKB1. It induces apoptosis in breast cancer cells, but not in other cancer cells, via a caspase-2 mediated pathway that involves mitochondrial membrane permeabilization but does not require other caspases. ITGB3BP may also act as an inhibitor of cyclin A-associated kinase. ITGB3BP may be involved in incorporation of newly synthesized CENPA into centromeres via its interaction with the CENPA-NAC complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: beta 3 endonexin; Beta-3-endonexin; beta3-endonexin; CENP-R; Centromere protein R; integrin beta 3 binding protein (beta3-endonexin); Integrin beta-3-binding protein; Nuclear receptor-interacting factor 3
Gene Aliases: CENP-R; CENPR; HSU37139; ITGB3BP; NRIF3; TAP20
UniProt ID: (Human) Q13352
Entrez Gene ID: (Human) 23421
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support