Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Keep as concentrated solution.
Predicted reactivity: Mouse (87%), Rat (88%), Pig (89%), Sheep (81%), Bovine (87%).
Positive Control: human plasma, rat plasma.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Ceruloplasmin encodes a protein that is a metalloprotein that binds most of the copper in plasma and is involved in the peroxidation of Fe(II)transferrin to Fe(III) transferrin. Mutations in this gene cause aceruloplasminemia, which results in iron accumulation and tissue damage, and is associated with diabetes and neurologic abnormalities.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bilitranslocase; Ceruloplasmin; Cuproxidase ceruloplasmin; Ferroxidase ceruloplasmin
Gene Aliases: CERP; CP; CP-2
UniProt ID: (Human) P00450
Entrez Gene ID: (Human) 1356, (Rat) 24268
Molecular Function:
oxidase
oxidoreductase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support