Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: MQESKAEGN NQAPGEEEEE EEEATNTHPP ASLPSQKYPG PQAEGDSEGL SQGLVDREKG LSAEPGWQAK REEEEEEEEE AEAGEEAVPE EEGPTVVLNP HPSLGYKEIR KGESRSEALA VDGAGKPGAE EAQDPEGKGE QEHSQQKEEE EEMAVVPQGL FRGGKSGELE QEEERLSKEW EDSKRWSKMD QLAKELTAEK RLEGQEEEED NRDSSMKLSF RARAYGFRGP GPQLRRGWRP SSREDSLEAG LPLQVRGYPE EKKEEEGSAN RRPEDQELES LSAIEAELEK VAHQLQALRR G (158-457 aa encoded by BC006459)
Chromogranin A (CHGA) is a member of the chromogranin/secretogranin family of neuroendocrine secretory proteins. It is found in secretory vesicles of neurons and endocine cells. CHGA is a precursor to three biologically active peptides: vasostatin, pancreastatin, and parastatin. These peptides act as autocine or paracrine negative modulators of the neuroendocrine system. CHGA is an excellent marker for carcinoid tumors, pheo-chromocytomas, paragangliomas, and other neuro-endocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: betagranin (N-terminal fragment of chromogranin A); catestatin; CgA; chromofungin; Chromogranin-A; CTNNB; DKFZp686D02253; FLJ25606; FLJ37923; MGC105435; parathyroid secretory protein 1; Pituitary secretory protein I; SP-I
Gene Aliases: CGA; CHGA; ChrA
UniProt ID: (Human) Q9BQB5, (Mouse) P26339
Entrez Gene ID: (Human) 1113, (Rat) 24258, (Mouse) 12652
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support